Learn More
enQuireBio™ Recombinant Human LZTFL1 Protein
A cDNA sequence encoding the LZTFL1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12612-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
LZTFL1 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
LZTFL1 protein solution (1 mg/ml) containing PBS buffer (pH 7.4), 10% glycerol and 1mM DTT. |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAELGLN EHHQNEVINY MRFARSKRGL RLKTVDSCFQ DLKESRLVED TFTIDEVSEV LNGLQAVVHS EVESELINTA YTNVLLLRQL FAQAEKWYLK LQTDISELEN RELLEQVAEF EKAEITSSNK KPILDVTKPK LAPLNEGGTA ELLNKEILRL QEENEKLKSR LKTIEIQATN ALDEKSKLEK ALQDLQLDQG NQKDFIKAQD LSNLENTVAA LKSEFQKTLN DKTENQKSLE ENLATAKHDL LRVQEQLHMA EKELEKKFQQ TAAYRNMKEI LTKKNDQIKD LRKRLAQYEP ED |