Learn More
enQuireBio™ Recombinant Human MAPRE1 Protein
A cDNA sequence encoding the MAPRE1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12644-20ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
MAPRE1 Protein | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MAPRE1 solution (0.5 mg/ml) 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 2mM DTT and 0.1M NaCl. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAVNVYSTSV TSDNLSRHDM LAWINESLQL NLTKIEQLCS GAAYCQFMDM LFPGSIALKK VKFQAKLEHE YIQNFKILQA GFKRMGVDKI IPVDKLVKGK FQDNFEFVQW FKKFFDANYD GKDYDPVAAR QGQETAVAPS LVAPALNKPK KPLTSSSAAP QRPISTQRTA AAPKAGPGVV RKNPGVGNGD DEAAELMQQV NVLKLTVEDL EKERDFYFGK LRNIELICQE NEGENDPVLQ RIVDILYATD EGFVIPDEGG PQEEQEEY |