missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MLLT11 Protein

A cDNA sequence encoding the MLLT11 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP12701-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15981269

  • 4985.00€ / 1mg
Expédition estimée: 10-07-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

MLLT11 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
MLLT11 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Greater than 80.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMRDPVSS QYSSFLFWRM PIPELDLSEL EGLGLSDTAT YKVKDSSVGK MIGQATAADQ EKNPEGDGLL EYSTFNFWRA PIASIHSFEL DLL
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions