missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MOSPD1 Protein

A cDNA sequence encoding the MOSPD1 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP12721-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15981329

  • 3740.00€ / 1mg
Expédition estimée: 17-07-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

MOSPD1 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
MOSPD1 protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4), 0.15M NaCl, 50% glycerol and 1mM DTT.
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMHQQKRQ PELVEGNLPV FVFPTELIFY ADDQSTHKQV LTLYNPYEFA LKFKVLCTTP NKYVVVDAAG AVKPQCCVDI VIRHRDVRSC HYGVIDKFRL QVSEQSQRKA LGRKEVVATL LPSAKEQQKE EEEKRLKEHL TESLFFEQSF QPENRAVSSG P
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions