Learn More
enQuireBio™ Recombinant Human MYL4 Protein
A cDNA sequence encoding the MYL4 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12769-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
MYL4 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MYL4 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 150mM NaCl, 1mM DTT and 20% glycerol. |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAPKKPEPKK EAAKPAPAPA PAPAPAPAPA PEAPKEPAFD PKSVKIDFTA DQIEEFKEAF SLFDRTPTGE MKITYGQCGD VLRALGQNPT NAEVLRVLGK PKPEEMNVKM LDFETFLPIL QHISRNKEQG TYEDFVEGLR VFDKESNGTV MGAELRHVLA TLGEKMTEAE VEQLLAGQED ANGCINYEAF VKHIMSGLEH HHHHH |