Learn More
enQuireBio™ Recombinant Human NDRG1 Protein
A cDNA sequence encoding the NDRG1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10791-50ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NDRG1 Protein | |
50 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MSREMQDVDL AEVKPLVEKG ETITGLLQEF DVQEQDIETL HGSVHVTLCG TPKGNRPVIL TYHDIGMNHK TCYNPLFNYE DMQEITQHFAVCHVDAPGQQ DGAASFPAGY MYPSMDQLAE MLPGVLQQFG LKSIIGMGTG AGAYILTRFA LNNPEMVEGL VLINVNPCAE GWMDWAASKI SGWTQALPDM VVSHLFGKEE MQSNVEVVHT YRQHIVNDMN PGNLHLFINA YNSRRDLEIE RPMPGTHTVT LQCPALLVVG DSSPAVDAVV ECNSKLDPTK TTLLKMADCG GLPQISQPAK LAEAFKYFVQ GMGYMPSASM TRLMRSRTAS GSSVTSLDGT RSRSHTSEGT RSRSHTSEGT RSRSHTSEGA HLDITPNSGA AGNSAGPKSM EVSCLEHHHH HH |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
Measured in a cell proliferation assay using MCF7 cell. The ED50 for this effect is 0.5 - 1.5ng/ml, corresponding to a Specific Activity of 666,000 -2,000,000 IU/mg. | |
Human | |
His | |
The NDRG1 solution contains 20mM Tris-HCl buffer (pH 8.0), 0.1mM PMSF and 10% Glycerol. |