Learn More
enQuireBio™ Recombinant Human NDUFA5 Protein
A cDNA sequence encoding the NDUFA5 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12812-100ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
4698 | |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
NDUFA5 | |
Human | |
His | |
The NDUFA5 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |
NDUFA5 Protein | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAGVLKK TTGLVGLAVC NTPHERLRIL YTKILDVLEE IPKNAAYRKY TEQITNEKLA MVKAEPDVKK LEDQLQGGQL EEVILQAEHE LNLARKMREW KLWEPLVEEP PADQWKWPI |