missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NEU1 Protein

A cDNA sequence encoding the NEU1 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP12830-5ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15901689

  • 131.00€ / 5µg
Expédition estimée: 28-06-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

NEU1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMENDFG LVQPLVTMEQ LLWVSGRQIG SVDTFRIPLI TATPRGTLLA FAEARKMSSS DEGAKFIALR RSMDQGSTWS PTAFIVNDGD VPDGLNLGAV VSDVETGVVF LFYSLCAHKA GCQVASTMLV WSKDDGVSWS TPRNLSLDIG TEVFAPGPGS GIQKQREPRK GRLIVCGHGT LERDGVFCLL SDDHGASWRY GSGVSGIPYG QPKQENDFNP DECQPYELPD GSVVINARNQ NNYHCHCRIV LRSYDACDTL RPRDVTFDPE LVDPVVAAGA VVTSSGIVFF SNPAHPEFRV NLTLRWSFSN GTSWRKETVQ LWPGPSGYSS LATLEGSMDG EEQAPQLYVL YEKGRNHYTE SISVAKISVY GTL
Greater than 85% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The NEU1 solution (0.25 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 10% glycerol.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions