Learn More
enQuireBio™ Recombinant Human NFE2L2 Protein
A cDNA sequence encoding the NFE2L2 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12834-2ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
4780 | |
Greater than 85.0% as determined by SDS-PAGE. | |
Research Use Only | |
NFE2L2 | |
Human | |
His | |
NFE2L2 protein solution (0.25 mg/ml) in Phosphate buffer saline (pH 7.4) containing 10% glycerol. |
NFE2L2 Protein | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MMDLELPPPG LPSQQDMDLI DILWRQDIDL GVSREVFDFS QRRKEYELEK QKKLEKERQE QLQKEQEKAF FAQLQLDEET GEFLPIQPAQ HIQSETSGSA NYSQVAHIPK SDALYFDDCM QLLAQTFPFV DDNEVSSATF QSLVPDIPGH IESPVFIATN QAQSPETSVA QVAPVDLDGM QQDIEQVWEE LLSIPELQCL NIENDKLVET TMVPSPEAKL TEVDNYHFYS SIPSMEKEVG NCSPHFLNAF EDSFSSILST EDPNQLTVNS LNSDATVNTD FGDEFYSAFI AEPSISNSMP SPATLSHSLS ELLNGPIDVS DLSLCKAFNQ NHPESTAEFN DSDSGISLNT SPSVASPEHS VESSSYGDTL LGLSDSEVEE LDSAPGSVKQ NGPKTPVHSS GDMVQPLSPS QGQSTHVHDA QCENTPEKEL PVSPGHRKTP FTKDKHSSRL EAHLTRDELR AKALHIPFPV EKIINLPVVD FNEMMSKEQF NEAQLALIRD IRRRGKNKVA AQNCRKRKLE NIVELEQDLD HLKDEKEKLL KEKGENDKSL HLLKKQLSTL YLEVFSMLRD EDGKPYSPSE YSLQQTRDGN VFLVPKSKKP DVKKN |