Learn More
enQuireBio™ Recombinant Human NMNAT2 Protein
A cDNA sequence encoding the NMNAT2 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10793-2ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
23057 | |
Greater than 85.0% as determined by SDS-PAGE. | |
Research Use Only | |
NMNAT2 | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MTETTKTHVI LLACGSFNPI TKGHIQMFER ARDYLHKTGR FIVIGGIVSP VHDSYGKQGL VSSRHRLIMC QLAVQNSDWI RVDPWECYQD TWQTTCSVLE HHRDLMKRVT GCILSNVNTP SMTPVIGQPQ NETPQPIYQN SNVATKPTAA KILGKVGESL SRICCVRPPV ERFTFVDENA NLGTVMRYEE IELRILLLCG SDLLESFCIP GLWNEADMEV IVGDFGIVVV PRDAADTDRI MNHSSILRKY KNNIMVVKDD INHPMSVVSS TKSRLALQHG DGHVVDYLSQ PVIDYILKSQ LYINASG |
NMNAT2 Protein | |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity: > 500 pmol/min/ug. One unit will convert 1.0 pmole of beta-NADH per minute to beta-NAD at PH 8.0 at 37C. | |
Human | |
His | |
NMNAT2 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |