missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human NOV/CCN3 Protein
Tous les produits Bio Techne Produits
Click to view available options
Quantité:
1 mg
100 μg
20 μg
5 μg
500 μg
Conditionnement:
100µg
1mg
20µg
500µg
5µg
Description
CCN3 Protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using murine Balb/c 3T3 cells is less than 1.0 μg/mL, corresponding to a specific activity of >1000 IU/mg.
Spécification
Spécification
| Numéro d’adhésion | P48745, P48745, P48745 |
| Concentration | LYOPH |
| À utiliser avec (application) | PAGE, Bioactivity |
| Formule | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM Tris-HCl, pH 8.6, 150 mM NaCl. |
| Identification génétique (Entrez) | 4856 |
| Poids moléculaire | 36.2 kDa |
| Quantité | 5 μg |
| Conditions de stockage | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Concentration en endotoxines | Less than 0.1 EU/ug of NOV/CCN3 as determined by LAL method. |
| Alias de gène | CCN family member 3, CCN3IGF-binding protein 9, IBP-9, IGFBP-9, IGFBP9novH, Insulin-like growth factor-binding protein 9, nephroblastoma overexpressed gene, Nephroblastoma-overexpressed gene protein homolog, NOVH, protein NOV homolog |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu