Learn More
enQuireBio™ Recombinant Human NUDCD2 Protein
A cDNA sequence encoding the NUDCD2 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12898-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NUDCD2 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NUDCD2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSAPFEE RSGVVPCGTP WGQWYQTLEE VFIEVQVPPG TRAQDIQCGL QSRHVALSVG GREILKGKLF DSTIADEGTW TLEDRKMVRI VLTKTKRDAA NCWTSLLESE YAADPWVQDQ MQRKLTLERF QKENPGFDFS GAEISGNYTK GGPDFSNLEK |