missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Optineurin / OPTN Protein
A DNA sequence encoding the Homo sapiens (Human) Optineurin / OPTN, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP9317-ye-100ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001008212.1 | |
Optineurin / OPTN Protein | |
Research Use Only | |
OPTN | |
Recombinant Protein | |
Yeast | |
MSHQPLSCLT EKEDSPSEST GNGPPHLAHP NLDTFTPEEL LQQMKELLTE NHQLKEAMKL NNQAMKGRFE ELSAWTEKQK EERQFFEIQS KEAKERLMAL SHENEKLKEE LGKLKGKSER SSEDPTDDSR LPRAEAEQEK DQLRTQVVRL QAEKADLLGI VSELQLKLNS SGSSEDSFVE IRMAEGEAEG SVKEIKHSPG PTRTVSTGTA LSKYRSRSAD GAKNYFEHEE LTVSQLLLCL REGNQKVERL EVALKEAKER VSDFEKKTSN RSEIETQTEG STEKENDEEK GPETVGSEVE ALNLQVTSLF KELQEAHTKL SKAELMKKRL QEKCQALERK NSAIPSELNE KQELVYTNKK LELQVESMLS EIKMEQAKTE DEKSKLTVLQ MTHNKLLQEH NNALKTIEEL TRKESEKVDR AVLKELSEKL ELAEKALASK QLQMDEMKQT IAKQEEDLET MTILRAQMEV YCSDFHAERA AREKIHEEKE QLALQLAVLL KENDAFEDGG RQSLMEMQSR HGARTSDSDQ QAYLVQRGAE DRDWRQQRNI PIHSCPKCGE VLPDIDTLQI HVMDCII | |
Greater than 90% as determined by SDS-PAGE. |
10133 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |