Learn More
enQuireBio™ Recombinant Human OXSM Protein
A cDNA sequence encoding the OXSM was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12943-2ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
OXSM Protein | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The OXSM solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 10% glycerol. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSKRKFFGT VPISRLHRRV VITGIGLVTP LGVGTHLVWD RLIGGESGIV SLVGEEYKSI PCSVAAYVPR GSDEGQFNEQ NFVSKSDIKS MSSPTIMAIG AAELAMKDSG WHPQSEADQV ATGVAIGMGM IPLEVVSETA LNFQTKGYNK VSPFFVPKIL VNMAAGQVSI RYKLKGPNHA VSTACTTGAH AVGDSFRFIA HGDADVMVAG GTDSCISPLS LAGFSRARAL STNSDPKLAC RPFHPKRDGF VMGEGAAVLV LEEYEHAVQR RARIYAEVLG YGLSGDAGHI TAPDPEGEGA LRCMAAALKD AGVQPEEISY INAHATSTPL GDAAENKAIK HLFKDHAYAL AVSSTKGATG HLLGAAGAVE AAFTTLACYY QKLPPTLNLD CSEPEFDLNY VPLKAQEWKT EKRFIGLTNS FGFGGTNATL CIAGL |