missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PAPSS1 Protein

A cDNA sequence encoding the PAPSS1 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP12958-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15982069

  • 5220.00€ / 1mg
Expédition estimée: 25-06-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

PAPSS1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMRATNV TYQAHHVSRN KRGQVVGTRG GFRGCTVWLT GLSGAGKTTV SMALEEYLVC HGIPCYTLDG DNIRQGLNKN LGFSPEDREE NVRRIAEVAK LFADAGLVCI TSFISPYTQD RNNARQIHEG ASLPFFEVFV DAPLHVCEQR DVKGLYKKAR AGEIKGFTGI DSEYEKPEAP ELVLKTDSCD VNDCVQQVVE LLQERDIVPV DASYEVKELY VPENKLHLAK TDAETLPALK INKVDMQWVQ VLAEGWATPL NGFMREREYL QCLHFDCLLD GGVINLSVPI VLTATHEDKE RLDGCTAFAL MYEGRRVAIL RNPEFFEHRK EERCARQWGT TCKNHPYIKM VMEQGDWLIG GDLQVLDRVY WNDGLDQYRL TPTELKQKFK DMNADAVFAF QLRNPVHNGH ALLMQDTHKQ LLERGYRRPV LLLHPLGGWT KDDDVPLMWR MKQHAAVLEE GVLNPETTVV AIFPSPMMYA GPTEVQWHCR ARMVAGANFY IVGRDPAGMP HPETGKDLYE PSHGAKVLTM APGLITLEIV PFRVAAYNKK KKRMDYYDSE HHEDFEFISG TRMRKLAREG QKPPEGFMAP KAWTVLTEYY KSLEKA
Greater than 85% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The PAPSS1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl and 20% glycerol.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions