Learn More
enQuireBio™ Recombinant Human PCP4L1 Protein
A cDNA sequence encoding the PCP4L1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12980-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
PCP4L1 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PCP4L1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 20% glycerol. |
Greater than 85.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSELNTK TSPATNQAAG QEEKGKAGNV KKAEEEEEID IDLTAPETEK AALAIQGKFR RFQKRKKDPS S |