Learn More
enQuireBio™ Recombinant Human Plastin-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Plastin-2, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP6289-ye-500ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_002289.2 | |
Plastin-2 Protein | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3936 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
LCP1 | |
Recombinant Protein | |
Yeast | |
ARGSVSDEEM MELREAFAKV DTDGNGYISF NELNDLFKAA CLPLPGYRVR EITENLMATG DLDQDGRISF DEFIKIFHGL KSTDVAKTFR KAINKKEGIC AIGGTSEQSS VGTQHSYSEE EKYAFVNWIN KALENDPDCR HVIPMNPNTN DLFNAVGDGI VLCKMINLSV PDTIDERTIN KKKLTPFTIQ ENLNLALNSA SAIGCHVVNI GAEDLKEGKP YLVLGLLWQV IKIGLFADIE LSRNEALIAL LREGESLEDL MKLSPEELLL RWANYHLENA GCNKIGNFST DIKDSKAYYH LLEQVAPKGD EEGVPAVVID MSGLREKDDI QRAECMLQQA ERLGCRQFVT ATDVVRGNPK LNLAFIANLF NRYPALHKPE NQDIDWGALE GETREERTFR NWMNSLGVNP RVNHLYSDLS DALVIFQLYE KIKVPVDWNR VNKPPYPKLG GNMKKLENCN YAVELGKNQA KFSLVGIGGQ DLNEGNRTLT LALIWQLMRR YTLNILEEIG GGQKVNDDII VNWVNETLRE AKKSSSISSF KDPKISTSLP VLDLIDAIQP GSINYDLLKT ENLNDDEKLN NAKYAISMAR KIGARVYALP EDLVEVNPKM VMTVFACLMG KGMKRV |