Learn More
enQuireBio™ Recombinant Human POLR2C Protein
A cDNA sequence encoding the POLR2C was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13096-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
POLR2C Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
POLR2C protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol 1mM DTT and 0.1M NaCl. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPYANQP TVRITELTDE NVKFIIENTD LAVANSIRRV FIAEVPIIAI DWVQIDANSS VLHDEFIAHR LGLIPLISDD IVDKLQYSRD CTCEEFCPEC SVEFTLDVRC NEDQTRHVTS RDLISNSPRV IPVTSRNRDN DPNDYVEQDD ILIVKLRKGQ ELRLRAYAKK GFGKEHAKWN PTAGVAFEYD PDNALRHTVY PKPEEWPKSE YSELDEDESQ APYDPNGKPE RFYYNVESCG SLRPETIVLS ALSGLKKKLS DLQTQLSHEI QSDVLTIN |