Learn More
enQuireBio™ Recombinant Human POLR2J2 Protein
A cDNA sequence encoding the POLR2J2 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13103-2ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
POLR2J2 Protein | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The POLR2J2 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol, 1mM DTT and 250mM Imidazole. |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMNAPPAF ESFLLFEGEK ITINKDTKVP KACLFTINKE DHTLGNIIKS QLLKDPQVLF AGYKVPHPLE HKIIIRVQTT PDYSPQEAFT NAITDLISEL SLLEERFRTC LLPLRLLP |