missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PPP4C Protein

A cDNA sequence encoding the PPP4C was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP13130-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15992589

  • 3915.00€ / 1mg
Expédition estimée: 10-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

PPP4C Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAEISDL DRQIEQLRRC ELIKESEVKA LCAKAREILV EESNVQRVDS PVTVCGDIHG QFYDLKELFR VGGDVPETNY LFMGDFVDRG FYSVETFLLL LALKVRYPDR ITLIRGNHES RQITQVYGFY DECLRKYGSV TVWRYCTEIF DYLSLSAIID GKIFCVHGGL SPSIQTLDQI RTIDRKQEVP HDGPMCDLLW SDPEDTTGWG VSPRGAGYLF GSDVVAQFNA ANDIDMICRA HQLVMEGYKW HFNETVLTVW SAPNYCYRCG NVAAILELDE HLQKDFIIFE AAPQETRGIP SKKPVADYFL
Greater than 90% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The PPP4C solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions