Learn More
enQuireBio™ Recombinant Human PTMS Protein
A cDNA sequence encoding the PTMS was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13207-10ug
Informations supplémentaires : Poids : 0.01000kg
Specifica
5763 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSEKSVE AAAELSAKDL KEKKEKVEEK ASRKERKKEV VEEEENGAEE EEEETAEDGE EEDEGEEEDE EEEEEDDEGP ALKRAAEEED EADPKRQKTE NGASA | |
Greater than 95.0% as determined by SDS-PAGE. |
PTMS Protein | |
Research Use Only | |
PTMS | |
Human | |
His | |
PTMS protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |