Learn More
enQuireBio™ Recombinant Human PTP4A3 Protein
A cDNA sequence encoding the PTP4A3 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13210-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
PTP4A3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MARMNRPAPV EVSYKHMRFL ITHNPTNATL STFIEDLKKY GATTVVRVCE VTYDKTPLEK DGITVVDWPF DDGAPPPGKV VEDWLSLVKA KFCEAPGSCV AVHCVAGLGR APVLVALALI ESGMKYEDAI QFIRQKRRGA INSKQLTYLE KYRPKQRLRFKDPHTHKTRC CVM | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PTP4A3 solution contains 20mM Tris-HCl buffer (pH 8.0), 2mM EDTA, 1mM DTT and 10% glycerol. |