missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RAB22A Protein

A cDNA sequence encoding the RAB22A was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP13233-1ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15982919

  • 125.00€ / 1µg
Expédition estimée: 18-07-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

RAB22A Protein
1 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
RAB22A protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 40% glycerol and 0.1M NaCl.
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MALRELKVCL LGDTGVGKSS IVWRFVEDSF DPNINPTIGA SFMTKTVQYQ NELHKFLIWD TAGQERFRAL APMYYRGSAA AIIVYDITKE ETFSTLKNWV KELRQHGPPN IVVAIAGNKC DLIDVREVME RDAKDYADSI HAIFVETSAK NAININELFI EISRRIPSTD ANLPSGGKGF KLRRQPSEPK RSCC
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions