missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RARA Protein

A cDNA sequence encoding the RARA was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP13269-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15993029

  • 4060.00€ / 1mg
Expédition estimée: 04-08-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

RARA Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The protein solution (1 mg/1ml) contains 20mM Tris-HCl pH 7.5, 0.1M NaCl and 5mM b-mercaptoethanol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MSEEIVPSPP SPPPLPRIYK PCFVCQDKSS GYHYGVSACE GCKGFFRRSI QKNMVYTCHR DKNCIINKVT RNRCQYCRLQ KCFEVGMSKESVRNDRNKKK KEVPKPE
Vidéos
FDS
Documents
Certificats
Promotions

Promotions