Learn More
enQuireBio™ Recombinant Human RCAN1 Protein
A cDNA sequence encoding the RCAN1 was constructed and used to recombinantly synthesize the protein.
Marque:  enQuireBio™ QP13286-1mg
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15923089
  • 5220.00€ / 1mg
Expédition estimée: 10-05-2024
Spécification
RCAN1 Protein
Research Use Only
Recombinant Protein
E. coli
MKLYFAQTLH IGSSHLAPPN PDKQFLISPP ASPPVGWKQV EDATPVINYD LLYAISKLGPGEKYELHAAT DTTPSVVVHV CESDQEKEEE EEMERMRRPK PKIIQTRRPE YTPIHLS
Greater than 90.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The protein solution (1 mg/ml) contains 20mM Tris-HCl pH 7.5, 100mM NaCl and 1mM DTT.