missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human REV1 Protein
A cDNA sequence encoding the REV1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13301-1mg
Additional Details : Poids : 0.01000kg
Specifications
REV1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGVAIYVNGY TDPSAEELRK LMMLHGGQYH VYYSRSKTTH IIATNLPNAK IKELKGEKVI RPEWIVESIK AGRLLSYIPY QLYTKQSSVQ KGLSFNPVCR PEDPLPGPSN IAKQLNNRVN HIVKKIETEN EVKVNGMNSW NEEDENNDFS FVDLEQTSPG RKQNGIPHPR GSTAIFNGHT PSSNGALKTQ DCLVPMVNSV ASRLSPA | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The REV1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |