Learn More
enQuireBio™ Recombinant Human RGS19 Protein
A cDNA sequence encoding the RGS19 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13310-5ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
RGS19 Protein | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RGS19 protein solution (0.5 mg/1ml) is formulated in Phosphate Buffered Saline (pH 7.4) and 10% Glycerol. |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MPTPHEAEKQ ITGPEEADRP PSMSSHDTAS PAAPSRNPCC LCWCCCCSCS WNQERRRAWQ ASRESKLQPL PSCEVCATPS PEEVQSWAQS FDKLMHSPAG RSVFRAFLRT EYSEENMLFW LACEELKAEA NQHVVDEKAR LIYEDYVSIL SPKEVSLDSR VREGINKKMQ EPSAHTFDDA QLQIYTLMHR DSYPRFLSSP TYRALLLQGP SQSSSEA |