Learn More
enQuireBio™ Recombinant Human RSPO3 Protein
A cDNA sequence encoding the RSPO3 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13366-5ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
RSPO3 Protein | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RSPO3 was lyophilized from a 0.2microm filtered solution in PBS, pH 7.4. |
Greater than 95% as determined by SDS PAGE. | |
Research Use Only | |
Recombinant Protein | |
HEK 293 | |
QNASRGRRQR RMHPNVSQGC QGGCATCSDY NGCLSCKPRL FFALERIGMK QIGVCLSSCP SGYYGTRYPD INKCTKCKAD CDTCFNKNFC TKCKSGFYLH LGKCLDNCPE GLEANNHTME CVSIVHCEVS EWNPWSPCTK KGKTCGFKRG TETRVREIIQ HPSAKGNLCP PTNETRKCTV DDIEGRMDEP KSCDKTHTCP PCPAPELLGG PSVFLFPPKP KDTLMISRTP EVTCVVVDVS HEDPEVKFNW YVDGVEVHNA KTKPREEQYN STYRVVSVLT VLHQDWLNGK EYKCKVSNKA LPAPIEKTIS KAKGQPREPQ VYTLPPSREE MTKNQVSLTC LVKGFYPSDI AVEWESNGQP ENNYKTTPPV LDSDGSFFLY SKLTVDKSRW QQGNVFSCSV MHEALHNHYT QKSLSLSPGK HHHHHH |