Learn More
enQuireBio™ Recombinant Human Selenide, water dikinase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Selenide, water dikinase 1, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP6663-ec-100ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001182531.1 | |
Selenide, water dikinase 1 Protein | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
22929 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
SEPHS1 | |
Recombinant Protein | |
E. coli | |
STRESFNPES YELDKSFRLT RFTELKGTGC KVPQDVLQKL LESLQENHFQ EDEQFLGAVM PRLGIGMDTC VIPLRHGGLS LVQTTDYIYP IVDDPYMMGR IACANVLSDL YAMGVTECDN MLMLLGVSNK MTDRERDKVM PLIIQGFKDA AEEAGTSVTG GQTVLNPWIV LGGVATTVCQ PNEFIMPDNA VPGDVLVLTK PLGTQVAVAV HQWLDIPEKW NKIKLVVTQE DVELAYQEAM MNMARLNRTA AGLMHTFNAH AATDITGFGI LGHAQNLAKQ QRNEVSFVIH NLPVLAKMAA VSKACGNMFG LMHGTCPETS GGLLICLPRE QAARFCAEIK SPKYGEGHQA WIIGIVEKGN RTARIIDKPR IIEVAPQVAT QNVNPTPGAT S |