missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Serpin F1/PEDF Protein
Tous les produits Bio Techne Produits
Click to view available options
Quantité:
100 μg
1 mg
20 μg
5 μg
500 μg
Conditionnement:
100µg
1mg
20µg
500µg
5µg
Description
PEDF Protein is fully biologically active when compared to standard. The ED50 as determined by its ability to enhance the adhesion of human Saos2 cells to bovine Collagen I coated plate is less than 2 ng/mL, corresponding to a specific activity of >5.0 x 10^5 IU/mg.
Spécification
Spécification
| Numéro d’adhésion | P36955, P36955, P36955 |
| À utiliser avec (application) | PAGE, Bioactivity |
| Formule | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, pH 7.4, 150 mM NaCl. |
| Identification génétique (Entrez) | 5176 |
| Poids moléculaire | M.W. (Theoretical): 44.4 kDa |
| Quantité | 500 μg |
| Conditions de stockage | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Concentration en endotoxines | Less than 1 EU/μg of Serpin F1/PEDF as determined by LAL method. |
| Alias de gène | Cell proliferation-inducing gene 35 protein, EPC-1PIG35, PEDFpigment epithelium-derived factor, pigment epithelium derived factor), member 1, proliferation-inducing protein 35, serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, Serpin F1, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 1 |
| Symbole de gène(s) | SERPINF1 |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu