missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SFRP4 Protein

A cDNA sequence encoding the SFRP4 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP13478-2ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15993699

  • 137.00€ / 2µg
Expédition estimée: 04-08-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

SFRP4 Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Flag Tag
Filtered (0.4microm) and lyophilized from 0.5 mg/ml in 0.05M phosphate buffer and 0.075M NaCl, pH 7.4.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
HEK 293
PGDYKDDDDK PAGVRGAPCE AVRIPMCRHM PWNITRMPNH LHHSTQENAI LAIEQYEELV DVNCSAVLRF FLCAMYAPIC TLEFLHDPIK PCKSVCQRAR DDCEPLMKMY NHSWPESLAC DELPVYDRGV CISPEAIVTD LPEDVKWIDI TPDMMVQERP LDVDCKRLSP DRCKCKKVKP TLATYLSKNY SYVIHAKIKA VQRSGCNEVT TVVDVKEIFK SSSPIPRTQV PLITNSSCQC PHILPHQDVL IMCYEWRSRM MLLENCLVEK WRDQLSKRSI QWEERLQEQR RTVQDKKKTA GRTSRSNPPK PKGKPPAPKP ASPKKNIKTR SAQKRTNPKR V
Vidéos
FDS
Documents
Certificats
Promotions

Promotions