Learn More
enQuireBio™ Recombinant Human SIRPG / SIRP gamma / CD172g Protein
A DNA sequence encoding the Homo sapiens (Human) SIRPG / SIRP gamma / CD172g, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP6687-ye-500ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001034597.1 | |
SIRPG / SIRP gamma / CD172g Protein | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
55423 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
SIRPG | |
Recombinant Protein | |
Yeast | |
EEELQMIQPE KLLLVTVGKT ATLHCTVTSL LPVGPVLWFR GVGPGRELIY NQKEGHFPRV TTVSDLTKRN NMDFSIRISS ITPADVGTYY CVKFRKGSPE NVEFKSGPGT EMALGAKPSA PVVLGPAART TPEHTVSFTC ESHGFSPRDI TLKWFKNGNE LSDFQTNVDP TGQSVAYSIR STARVVLDPW DVRSQVICEV AHVTLQGDPL RGTANLSEAI RVPPTLEVTQ QPMRVGNQVN VTCQVRKFYP QSLQLTWSEN GNVCQRETAS TLTENKDGTY NWTSWFLVNI SDQRDDVVLT CQVKHDGQLA VSKRLALEVT VHQKDQSSDA TP |