Learn More
enQuireBio™ Recombinant Human SLPI Protein
A cDNA sequence encoding the SLPI was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13524-2ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
SLPI Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSGKSFKAGV CPPKKSAQCL RYKKPECQSD WQCPGKKRCC PDTCGIKCLD PVDTPNPTRR KPGKCPVTYG QCLMLNPPNF CEMDGQCKRD LKCCMGMCGK SCVSPVKA | |
Greater than 80.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SLPI Human solution containing 20mM Tris-HCl pH 8.0, 2mM DTT, 0.1M NaCl and 20% glycerol. |