Learn More
enQuireBio™ Recombinant Human SLPI Protein
A cDNA sequence encoding the SLPI was constructed and used to recombinantly synthesize the protein.
Marque:  enQuireBio™ QP13524-10ug
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15973849
  • 203.00€ / 10µg
Expédition estimée: 25-06-2024
Spécification
SLPI Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MSGKSFKAGV CPPKKSAQCL RYKKPECQSD WQCPGKKRCC PDTCGIKCLD PVDTPNPTRR KPGKCPVTYG QCLMLNPPNF CEMDGQCKRD LKCCMGMCGK SCVSPVKA
Greater than 80.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
SLPI Human solution containing 20mM Tris-HCl pH 8.0, 2mM DTT, 0.1M NaCl and 20% glycerol.