Learn More
enQuireBio™ Recombinant Human SNAP23 Protein
A cDNA sequence encoding the SNAP23 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13531-25ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
SNAP23 Protein | |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The protein solution contains 20mM Tris-HCl pH 8. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MDNLSSEEIQ QRAHQITDES LESTRRILGL AIESQDAGIK TITMLDEQKE QLNRIEEGLD QINKDMRETE KTLTELNKCC GLCVCPCNRT KNFESGKAYK TTWGDGGENS PCNVVSKQPG PVTNGQLQQP TTGAASGGYI KRITNDARED EMEENLTQVG SILGNLKDMA LNIGNEIDAQ NPQIKRITDK ADTNRDRIDI ANARAKKLID S |