missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Spermatogenesis-defective protein 39 homolog Protein
A DNA sequence encoding the Homo sapiens (Human) Spermatogenesis-defective protein 39 homolog, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP8216-ec-10ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
NP_001180243.1 | |
Spermatogenesis-defective protein 39 homolog Protein | |
Research Use Only | |
VIPAS39 | |
Recombinant Protein | |
E. coli | |
MNRTKGDEEE YWNSSKFKAF TFDDEDDELS QLKESKRAVN SLRDFVDDDD DDDLERVSWS GEPVGSISWS IRETAGNSGS THEGREQLKS RNSFSSYAQL PKPTSTYSLS SFFRGRTRPG SFQSLSDALS DTPAKSYAPE LGRPKGEYRD YSNDWSPSDT VRRLRKGKVC SLERFRSLQD KLQLLEEAVS MHDGNVITAV LIFLKRTLSK EILFRELEVR QVALRHLIHF LKEIGDQKLL LDLFRFLDRT EELALSHYRE HLNIQDPDKR KEFLKTCVGL PFSAEDSAHI QDHYTLLERQ IIIEANDRHL ESAGQTEIFR KHPRKASILN MPLVTTLFYS CFYHYTEAEG TFSSPVNLKK TFKIPDKQYV LTALAARAKL RAWNDVDALF TTKNWLGYTK KRAPIGFHRV VEILHKNNAP VQILQEYVNL VEDVDTKLNL ATKFKCHDVV IDTYRDLKDR QQLLAYRSKV DKGSAEEEKI DALLSSSQIR WKN | |
Greater than 90% as determined by SDS-PAGE. |
63894 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |