Learn More
enQuireBio™ Recombinant Human SPINK7 Protein
A cDNA sequence encoding the SPINK7 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13576-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
SPINK7 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SPINK7 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 40% glycerol and 2mM DTT. |
Greater than 85.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSSEAASLS PKKVDCSIYK KYPVVAIPCP ITYLPVCGSD YITYGNECHL CTESLKSNGR VQFLHDGSC |