missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Splicing factor U2AF 35 kDa subunit Protein
A DNA sequence encoding the Homo sapiens (Human) Splicing factor U2AF 35 kDa subunit, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP6861-ec-200ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001020374.1 | |
Splicing factor U2AF 35 kDa subunit Protein | |
Research Use Only | |
U2AF1 | |
Recombinant Protein | |
E. coli | |
MAEYLASIFG TEKDKVNCSF YFKIGACRHG DRCSRLHNKP TFSQTIALLN IYRNPQNSSQ SADGLRCAVS DVEMQEHYDE FFEEVFTEME EKYGEVEEMN VCDNLGDHLV GNVYVKFRRE EDAEKAVIDL NNRWFNGQPI HAELSPVTDF REACCRQYEM GECTRGGFCN FMHLKPISRE LRRELYGRRR KKHRSRSRSR ERRSRSRDRG RGGGGGGGGG GGGRERDRRR SRDRERSGRF | |
Greater than 80% as determined by SDS-PAGE. |
102724594 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |