Learn More
enQuireBio™ Recombinant Human TGF-beta 1 / TGFB1 Protein
Recombinant Protein
Marque:  enQuireBio™ QP773-500ug
Informations supplémentaires : Poids : 0.01000kg
Code produit. 16000481
  • 4665.00€ / 500µg
Expédition estimée: 09-08-2022
Spécification
NP_000651.3
TGF-beta 1 / TGFB1 Protein
500 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity at a concentration of 500 pg/mL in the Inhibition of IL-4-induced HT-2 cell proliferation assay. The bioactivity acceptance criteria is set at 2 x 10^6 units/mg.
Human
Untagged
0.1 % trifluoroacetic acid (TFA) and Trehalose in a 20:1 Trehalose to protein ratio
7040
Greater than 95% as determined by SDS-PAGE.
Research Use Only
TGFB1
Recombinant Protein
HO cells
ALDTNYCFSS TEKNCCVRQL YIDFRKDLGW KWIHEPKGYH ANFCLGPCPY IWSLDTQYSK VLALYNQHNP GASAAPCCVP QALEPLPIVY YVGRKPKVEQ LSNMIVRSCK CS