missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thyroglobulin Protein
A cDNA sequence encoding the Thyroglobulin was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13748-20ug
Additional Details : Poids : 0.01000kg
Specifications
Thyroglobulin Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
MALVLEIFTL LASICWVSAN IFEYQVDAQP LRPCELQRET AFLKQADYVP QCAEDGSFQT VQCQNDGRSC WCVGANGSEV LGSRQPGRPV ACLSFCQLQK QQILLSGYIN STDTSYLPQC QDSGDYAPVQ CDVQQVQCWC VDAEGMEVYG TRQLGRPKRC PRSCEIRNRR LLHGVGDKSP PQCSAEGEFM PVQCKFVNTT DMMIFDLVHS YNRFPDAFVT FSSFQRRFPE VSGYCHCADS QGRELAETGL ELLLDEIYDT IFAGLDLPST FTETTLYRIL QRRFLAVQSV ISGRFRCPTK CEVERFTATS FGHPYVPSCR RNGDYQAVQC QTEGPCWCVD AQGKEMHGTR QQGEPPSCAE GQSCASERQQ ALSRLYFGTS GYFSQHDLFS SPEKRWASPR VARFATSCPP TIKELFVDSG LLRPMVEGQS QQFSVSENLL KEAIRAIFPS RGLARLALQF TTNPKRLQQN LFGGKFLVNV GQFNLSGALG TRGTFNFSQF FQQLGLASFL NGGRQEDLAK PLSVGLDSNS STGTPEAAKK DGTMNKPTVG SFGFEINLQE NQNALKFLAS LLELPEFLLF LQHAISVPED VARDLGDVME TVLSSQTCEQ TPERLFVPSC TTEGSYEDVQ CFSGECWCVN SWGKELPGSR VRGGQPRCPT DCEKQRARMQ SLMGSQPAGS TLFVPACTSE GHFLPVQCFN SECYCVDAEG QAIPGTRSAI GKPKKCPTPC QLQSEQAFLR TVQALLSNSS MLPTLSDTYI PQCSTDGQWR QVQCNGPPEQ VFELYQRWEA QNKGQDLTPA KLLVKIMSYR EAASGNFSLF IQSLYEAGQQ DVFPVLSQYP SLQDVPLAAL EGKRPQPREN ILLEPYLFWQ ILNGQLSQYP GSYSDFSTPL AHFDLRNCWC VDEAGQELEG | |
Greater than 95% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Human Thyroglobulin is supplied in 20mM HEPES buffer pH 7.6, 150mM NaCl and 40% Sucrose (w/v). |