Learn More
enQuireBio™ Recombinant Human Transcription initiation factor IIB Protein
A DNA sequence encoding the Homo sapiens (Human) Transcription initiation factor IIB, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP6130-ec-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001505.1 | |
Transcription initiation factor IIB Protein | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
2959 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
GTF2B | |
Recombinant Protein | |
E. coli | |
MASTSRLDAL PRVTCPNHPD AILVEDYRAG DMICPECGLV VGDRVIDVGS EWRTFSNDKA TKDPSRVGDS QNPLLSDGDL STMIGKGTGA ASFDEFGNSK YQNRRTMSSS DRAMMNAFKE ITTMADRINL PRNIVDRTNN LFKQVYEQKS LKGRANDAIA SACLYIACRQ EGVPRTFKEI CAVSRISKKE IGRCFKLILK ALETSVDLIT TGDFMSRFCS NLCLPKQVQM AATHIARKAV ELDLVPGRSP ISVAAAAIYM ASQASAEKRT QKEIGDIAGV ADVTIRQSYR LIYPRAPDLF PTDFKFDTPV DKLPQL |