Learn More
enQuireBio™ Recombinant Human Tropomodulin-4 Protein
A DNA sequence encoding the Homo sapiens (Human) Tropomodulin-4, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP7523-ec-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_037485.2 | |
Tropomodulin-4 Protein | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
29765 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
TMOD4 | |
Recombinant Protein | |
E. coli | |
MSSYQKELEK YRDIDEDEIL RTLSPEELEQ LDCELQEMDP ENMLLPAGLR QRDQTKKSPT GPLDREALLQ YLEQQALEVK ERDDLVPFTG EKKGKPYIQP KREIPAEEQI TLEPELEEAL AHATDAEMCD IAAILDMYTL MSNKQYYDAL CSGEICNTEG ISSVVQPDKY KPVPDEPPNP TNIEEILKRV RSNDKELEEV NLNNIQDIPI PMLSELCEAM KANTYVRSFS LVATRSGDPI ANAVADMLRE NRSLQSLNIE SNFISSTGLM AVLKAVRENA TLTELRVDNQ RQWPGDAVEM EMATVLEQCP SIVRFGYHFT QQGPRARAAQ AMTRNNELRR QQKKR |