Learn More
enQuireBio™ Recombinant Human TSSK2 Protein
A cDNA sequence encoding the TSSK2 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13832-2ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
23617 | |
Greater than 85% as determined by SDS-PAGE. | |
Research Use Only | |
TSSK2 | |
Human | |
His | |
TSSK2 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |
TSSK2 Protein | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDDATVL RKKGYIVGIN LGKGSYAKVK SAYSERLKFN VAVKIIDRKK TPTDFVERFL PREMDILATV NHGSIIKTYE IFETSDGRIY IIMELGVQGD LLEFIKCQGA LHEDVARKMF RQLSSAVKYC HDLDIVHRDL KCENLLLDKD FNIKLSDFGF SKRCLRDSNG RIILSKTFCG SAAYAAPEVL QSIPYQPKVY DIWSLGVILY IMVCGSMPYD DSDIRKMLRI QKEHRVDFPR SKNLTCECKD LIYRMLQPDV SQRLHIDEIL SHSWLQPPKP KATSSASFKR EGEGKYRAEC KLDTKTGLRP DHRPDHKLGA KTQHRLLVVP ENENRMEDRL AETSRAKDHH ISGAEVGKAS T |