Learn More
enQuireBio™ Recombinant Human UBE2D1 Protein
A cDNA sequence encoding the UBE2D1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13855-20ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
UBE2D1 Protein | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
UBE2D1 solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 40% glycerol and 0.1M NaCl. |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMALKRIQ KELSDLQRDP PAHCSAGPVG DDLFHWQATI MGPPDSAYQG GVFFLTVHFP TDYPFKPPKI AFTTKIYHPN INSNGSICLD ILRSQWSPAL TVSKVLLSIC SLLCDPNPDD PLVPDIAQIY KSDKEKYNRH AREWTQKYAM |