missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ULBP3 Protein

A cDNA sequence encoding the ULBP3 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP13887-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15995019

  • 7815.00€ / 1mg
Expédition estimée: 04-08-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

ULBP3 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
ULBP3 was filtered (0.4 microm) and lyophilized in 20mM Tris buffer, 50mM NaCl and 0.1% v/v amisoft CS-22, pH 7.5.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHASDAHSLWYNFT IIHLPRHGQQ WCEVQSQVDQ KNFLSYDCGS DKVLSMGHLE EQLYATDAWG KQLEMLREVG QRLRLELADT ELEDFTPSGP LTLQVRMSCE CEADGYIRGS WQFSFDGRKF LLFDSNNRKW TVVHAGARRM KEKWEKDSGL TTFFKMVSMR DCKSWLRDFL MHRKKRLEPT APPTMAPG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions