missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VAPA Protein
A cDNA sequence encoding the VAPA was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13921-25ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
VAPA Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
RGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMAS ASGAMAKHEQ ILVLDPPTDL KFKGPFTDVV TTNLKLRNPS DRKVCFKVKT TAPRRYCVRP NSGIIDPGST VTVSVMLQPF DYDPNEKSKH KFMVQTIFAP PNTSDMEAVW KEAKPDELMD SKLRCVFEMP NENDKLNDME PSKAVPLNAS KQDGPMPKPH SVSLNDTETR KLMEECKRLQ GEMMKLSEEN RHLRDEGLRL RKVAHSDKPG STSTASFRDN VTSP | |
Greater than 85% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
VAPA solution containing 20mM Tris pH 8.0, 1mM DTT and 10% glycerol. |