Learn More
enQuireBio™ Recombinant Human WWTR1 Protein
A cDNA sequence encoding the WWTR1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13968-20ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
WWTR1 Protein | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The WWTR1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol. |
Greater than 85.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMNPASAP PPLPPPGQQV IHVTQDLDTD LEALFNSVMN PKPSSWRKKI LPESFFKEPD SGSHSRQSST DSSGGHPGPR LAGGAQHVRS HSSPASLQLG TGAGAAGSPA QQHAHLRQQS YDVTDELPLP PGWEMTFTAT GQRYFLNHIE KITTWQDPRK AMNQPLNHMN LHPAVSSTPV PQRSMAVSQP NLVMNHQHQQ QMAPSTLSQQ NHPTQNPPAG LMSMPNALTT QQQQQQKLRL QRIQMERERI RMRQEELMRQ EAALCRQLPM EAETLAPVQA AVNPPTMTPD MRSITNNSSD PFLNGGPYHS REQSTDSGLG LGCYSVPTTP EDFLSNVDEM DTGENAGQTP MNINPQQTRF PDFLDCLPGT NVDLGTLESE DLIPLFNDVE SALNKSEPFL TWL |