Learn More
enQuireBio™ Recombinant M. tuberculosis ESAT-6-like protein EsxH Protein
A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) ESAT-6-like protein EsxH, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP7384-ec-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
WP_003401514.1 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
esxH | |
Recombinant Protein | |
E. coli | |
SQIMYNYPAM LGHAGDMAGY AGTLQSLGAE IAVEQAALQS AWQGDTGITY QAWQAQWNQA MEDLVRAYHA MSSTHEANTM AMMARDTAEA AKWGG |
M. tuberculosis ESAT-6-like protein EsxH Protein | |
1 mg | |
Not determined. | |
Full Length | |
M. tuberculosis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |