Learn More
enQuireBio™ Recombinant Mouse Acetylcholine receptor subunit alpha Protein
A DNA sequence encoding the Mus musculus (Mouse) Acetylcholine receptor subunit alpha, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5833-ye-100ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_031415.2 | |
Acetylcholine receptor subunit alpha Protein | |
100 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
11435 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
Chrna1 | |
Recombinant Protein | |
Yeast | |
SEHETRLVAK LFEDYSSVVR PVEDHREIVQ VTVGLQLIQL INVDEVNQIV TTNVRLKQQW VDYNLKWNPD DYGGVKKIHI PSEKIWRPDV VLYNNADGDF AIVKFTKVLL DYTGHITWTP PAIFKSYCEI IVTHFPFDEQ NCSMKLGTWT YDGSVVAINP ESDQPDLSNF MESGEWVIKE ARGWKHWVFY SCCPTTPYLD ITYHFVMQRL |