missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Arginase 1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Arginase 1, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP7724-ec-50ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_031508.1 | |
Arginase 1 Protein | |
Research Use Only | |
ARG1 | |
Recombinant Protein | |
E. coli | |
MSSKPKSLEI IGAPFSKGQP RGGVEKGPAA LRKAGLLEKL KETEYDVRDH GDLAFVDVPN DSSFQIVKNP RSVGKANEEL AGVVAEVQKN GRVSVVLGGD HSLAVGSISG HARVHPDLCV IWVDAHTDIN TPLTTSSGNL HGQPVSFLLK ELKGKFPDVP GFSWVTPCIS AKDIVYIGLR DVDPGEHYII KTLGIKYFSM TEVDKLGIGK VMEETFSYLL GRKKRPIHLS FDVDGLDPAF TPATGTPVLG GLSYREGLYI TEEIYKTGLL SGLDIMEVNP TLGKTAEEVK STVNTAVALT LACFGTQREG NHKPGTDYLK PPK | |
Greater than 90% as determined by SDS-PAGE. |
11846 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |